Learn More
Torsemide, MedChemExpress
Product Code. 16488186
Encompass_Preferred
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
Product Code. 16488186
Brand: Medchem Express HYB0247
MedChemExpress Torsemide (Torasemide) is an orally active loop diuretic. Torsemide has anti-aldosterone and vasodilatory effects. Torsemide also can be used for the research of heart failure, renal disease and hepatic cirrhosis.
Torsemide (Torasemide) is an orally active loop diuretic. Torsemide has anti-aldosterone and vasodilatory effects. Torsemide also can be used for the research of heart failure, renal disease and hepatic cirrhosis.
Specifications
Chemical Name or Material Torsemide
CAS 56211-40-6
Color White
Purity Grade Notes Research
Molecular Formula C16H20N4O3S
Quantity 100 mg
Synonym Torasemide
Solubility Information DMSO : ≥ 3.5 mg/mL (10.05 mM)
SMILES O=S(C1=C(NC2=CC=CC(C)=C2)C=CN=C1)(NC(NC(C)C)=O)=O
Molecular Weight (g/mol) 348.42
Formula Weight 348.42
Percent Purity 99.85%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.