Learn More
Topotecan-d6 Carboxylic Acid Sodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Topotecan-d6 Carboxylic Acid Sodium Salt |
| CAS | 1263470-27-4 |
| Molecular Formula | C23 D6 H18 N3 Na O6 |
| Quantity | 1 mg |
| InChI Formula | InChI=1S/C23H25N3O6.Na/c1-4-23(32,22(30)31)16-8-18-20-12(9-26(18)21(29)15(16)11-27)7-13-14(10-25(2)3)19(28)6-5-17(13)24-20;/h5-8,27-28,32H,4,9-11H2,1-3H3,(H,30,31);/q;+1/p-1/i2D3,3D3; |
| SMILES | [2H]C([2H])([2H])N(Cc1c(O)ccc2nc3C4=CC(=C(CO)C(=O)N4Cc3cc12)C(O)(CC)C(=O)O[Na])C([2H])([2H])[2H] |
| IUPAC Name | [2-[1-[[bis(trideuteriomethyl)amino]methyl]-2-hydroxy-8-(hydroxymethyl)-9-oxo-11H-indolizino[1,2-b]quinolin-7-yl]-2-hydroxybutanoyl]oxysodium |
| Molecular Weight (g/mol) | 467.48 |
| Formula Weight | 467.194 |