missing translation for 'onlineSavingsMsg'
Learn More

Topiramate-13C6, TRC

Product Code. 30455884
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30455884 5 mg 5mg
30451508 1 mg 1mg
30453331 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30455884 Supplier Toronto Research Chemicals Supplier No. TRCT5402515MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula 13C6 C6 H21 N O8 S
Molecular Weight (g/mol) 345.318
Synonym 2,3:4,5-Bis-O-(1-methylethylidene)-beta-D-fuctopyranose 1-Sulfamate-13C6, Epitoma-13C6, Epitomax-13C6, McN 4853-13C6, RWJ 17021-13C6, TPM-13C6, Topamac-13C6, Topamax-13C6, Topimax-13C6, Topina-13C6, Topomax-13C6
IUPAC Name [(1R,2S,6S,9R)-4,4,11,11-tetra((113C)methyl)-3,5,7,10,12-pentaoxatricyclo[7.3.0.02,6]dodecan-6-yl]methyl sulfamate
SMILES [13CH3][13C]1([13CH3])O[C@@H]2CO[C@@]3(COS(=O)(=O)N)O[13C]([13CH3])([13CH3])O[C@H]3[C@@H]2O1

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C12H21NO8S/c1-10(2)18-7-5-16-12(6-17-22(13,14)15)9(8(7)19-10)20-11(3,4)21-12/h7-9H,5-6H2,1-4H3,(H2,13,14,15)/t7-,8-,9+,12+/m1/s1/i1+1,2+1,3+1,4+1,10+1,11+1
Formula Weight 345.119
Percent Purity >95%
Chemical Name or Material Topiramate-13C6
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.