missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tool Steel, AISI O6, T31506, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Tool Steel, AISI O6, T31506, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Aluminium/Arsenic/Boron/Carbon/Chromium/Cobalt/Copper/Lead/Manganese/Molybdenum/Nickel/Niobium/Nitrogen/Oxygen/Phosphorus/Silicon/Tin/Titanium/Vanadium |
| CAS | 7439-89-6 |
| Form | Disc |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction