Learn More
TOMM5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25607-1-AP-150UL
 View more versions of this product
Product Code. 16834175
  • 4836.10 NOK / 150µL
Description
TOMM5, also named as C9orf105 and TOM5, has three isoforms with MW 6 kDa and 9-10 kDa.
Specifications
Specifications
TOMM5
Polyclonal
Unconjugated
TOMM5
C9orf105, RP11 263I4.1, Tom5, TOMM5
Rabbit
Antigen Affinity Chromatography
RUO
401505, 68512
-20°C
Liquid
Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
B1AXP6, Q8N4H5
TOMM5
TOMM5 Fusion Protein Ag22275
150 μL
Primary
Human, Mouse