missing translation for 'onlineSavingsMsg'
Learn More

TOMM5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 25607-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16844175

  • 1415.13 NOK / 20µL
Estimated Shipment: 10-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

TOMM5, also named as C9orf105 and TOM5, has three isoforms with MW 6 kDa and 9-10 kDa.
Specifications

Specifications

TOMM5
Polyclonal
Unconjugated
TOMM5
C9orf105, RP11 263I4.1, Tom5, TOMM5
Rabbit
Antigen Affinity Chromatography
RUO
401505, 68512
-20°C
Liquid
Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
B1AXP6, Q8N4H5
TOMM5
TOMM5 Fusion Protein Ag22275
20 μL
Primary
Mouse, Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers