Learn More
TOM20 Rabbit anti-Human, Unconjugated, Clone: 1D22, Proteintech
Rabbit Recombinant Antibody
Brand:  Proteintech 80501-1-RR-100UL
 View more versions of this product
Product Code. 17561506
  • 5178.17 NOK / 100µL
Description
The TOM20 antibody from Proteintech is a rabbit recombinant antibody to a recombinant protein of human TOM20. This antibody recognizes human, mouse, rat, pig, chicken antigen. The TOM20 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
TOM20
1D22
Rabbit
Primary
Human, Mouse, Rat, Pig, Chicken
Western Blot, Immunohistochemistry
Unconjugated
100 μL
9804