missing translation for 'onlineSavingsMsg'
Learn More

Toluene-4-sulfonic acid monohydrate, for analysis EMSURE™, ACS, MilliporeSigma™

Product Code. 15885350
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15885350 500 g Plastic Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15885350 Supplier Supelco Supplier No. 1096130500
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 56°C
CAS 6192-52-5
Quantity 500 g
Packaging Plastic Bottle
Molecular Formula C7H10O4S
UN Number 2585
Synonym p-toluenesulfonic acid monohydrate, 4-methylbenzenesulfonic acid hydrate, 4-methylbenzenesulfonic acid monohydrate, p-toluene sulfonic acid monohydrate, unii-3bto78gaff, benzenesulfonic acid, 4-methyl-, monohydrate, ptoluenesulfonic acid, p-toluenesulfonic acid, monohydrate, 3bto78gaff, 4-methylbenzene-1-sulfonic acid hydrate
Boiling Point 140°C
InChI Key KJIFKLIQANRMOU-UHFFFAOYSA-N
SMILES CC1=CC=C(C=C1)S(=O)(=O)O.O
IUPAC Name 4-methylbenzenesulfonic acid;hydrate
Molecular Weight (g/mol) 190.213
PubChem CID 521998
Grade ACS
Chemical Name or Material Toluene-4-sulfonic Acid Monohydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.