Learn More
Toltrazuril, TRC
Product Code. 30135796
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135796 1 g 1g
30138074 250 mg 250mg
17944155 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135796 Supplier Toronto Research Chemicals Supplier No. TRCT5358501G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 69004-03-1
Molecular Formula C18 H14 F3 N3 O4 S
Molecular Weight (g/mol) 425.38
Synonym 1-Methyl-3-[3-methyl-4-[4-[(trifluoromethyl)thio]phenoxy]phenyl]-1,3,5-triazine-2,4,6(1H,3H,5H)-trione, BAY-Vi 9142, Baycox, Deltazuril, Toltrazuril, 1,3,5-Triazine-2,4,6(1H,3H,5H)-trione, 1-methyl-3-[3-methyl-4-[4-[(trifluoromethyl)thio]phenoxy]phenyl]-
IUPAC Name 1-methyl-3-[3-methyl-4-[4-(trifluoromethylsulfanyl)phenoxy]phenyl]-1,3,5-triazinane-2,4,6-trione
SMILES CN1C(=O)NC(=O)N(C1=O)c2ccc(Oc3ccc(SC(F)(F)F)cc3)c(C)c2
Specifications
Quantity 1 g
InChI Formula InChI=1S/C18H14F3N3O4S/c1-10-9-11(24-16(26)22-15(25)23(2)17(24)27)3-8-14(10)28-12-4-6-13(7-5-12)29-18(19,20)21/h3-9H,1-2H3,(H,22,25,26)
Formula Weight 425.0657
Percent Purity >95%
Chemical Name or Material Toltrazuril
Grade HPLC