missing translation for 'onlineSavingsMsg'
Learn More

Toltrazuril (sulfone), MedChemExpress

Product Code. p-200099639
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. Quantity unitSize
16463277 50 mg 50mg
16407268 10 mg 10mg
2 options
This item is not returnable. View return policy

Product Code. 16463277

Brand: Medchem Express HY17008

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Toltrazuril sulfone (Ponazuril) is a metabolite of Toltrazuril (HY-B0175), with antiprotozoal activity. Toltrazuril sulfone is a triazine anticoccidial that is developed to prevent coccidiosis in poultry.

Toltrazuril sulfone (Ponazuril) is a metabolite of Toltrazuril (HY-B0175), with antiprotozoal activity. Toltrazuril sulfone is a triazine anticoccidial that is developed to prevent coccidiosis in poultry.

Specifications

Chemical Name or Material Toltrazuril (sulfone)
Physical Form Solid
CAS 69004-04-2
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C18H14F3N3O6S
For Use With (Application) COVID-19-immunoregulation
Synonym Ponazuril
Solubility Information DMSO : 50 mg/mL (109.32 mM; Need ultrasonic)
SMILES O=S(C1=CC=C(C=C1)OC2=C(C=C(C=C2)N3C(N(C(NC3=O)=O)C)=O)C)(C(F)(F)F)=O
Molecular Weight (g/mol) 457.38
Formula Weight 457.38
Percent Purity 99.34%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.