Learn More
Toloxatone, MedChemExpress
Product Code. 16498607
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16498607
Brand: Medchem Express HY14196
MedChemExpress Toloxatone (MD 69276) is a reversible monoamine oxidase A (MAOA) inhibitor. Antidepressant.
Toloxatone (MD 69276) is a reversible monoamine oxidase A (MAOA) inhibitor. Antidepressant.
Specifications
Chemical Name or Material Toloxatone
Physical Form Solid
CAS 29218-27-7
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C11H13NO3
For Use With (Application) Neuroscience-Neuromodulation
Synonym MD 69276
Solubility Information DMSO : 125 mg/mL (603.19 mM; Need ultrasonic)
SMILES CC1=CC(N2C(OC(CO)C2)=O)=CC=C1
Molecular Weight (g/mol) 207.23
Formula Weight 207.23
Percent Purity 99.84%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.