Learn More
Tolazoline hydrochloride, MedChemExpress
Product Code. 16470718
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16470718 5 g 5g
16432009 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16470718 Supplier Medchem Express Supplier No. HYA0066A
This item requires a price update prior to being ordered.
Request a Price for this item here.
, 02 950 59 478
MedChemExpress Tolazoline (hydrochloride)(Imidaline (hydrochloride)) Hcl is a non-selective competitive α-adrenergic receptor antagonist.
Tolazoline (hydrochloride)(Imidaline (hydrochloride)) Hcl is a non-selective competitive α-adrenergic receptor antagonist.
Specifications
Chemical Name or Material Tolazoline hydrochloride
Physical Form Solid
CAS 59-97-2
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C10H13ClN2
Synonym Imidaline hydrochlorideNSC35110 hydrochloride
Solubility Information H2O : ≥ 50 mg/mL (254.22 mM) ∣DMSO : 33.33 mg/mL (169.46 mM; Need ultrasonic)
SMILES [H]Cl.C1(CC2=CC=CC=C2)=NCCN1
Molecular Weight (g/mol) 196.68
Formula Weight 196.68
Percent Purity 98.0%
Grade Research