Description
The TOE1 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human TOE1. This antibody recognizes human antigen. The TOE1 antibody has been validated for the following applications: WB,ELISA analysis.
Specifications
Specifications
| Antigen | TOE1 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 2C10H7 |
| Concentration | 1000 μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Quantity | 20 μL |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 114034 |
| Show More |