Learn More
Tocris Bioscience™ 2-Furoyl-LIGRLO-amide
GREENER_CHOICE
Product Code. 18075272
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. Quantity unitSize
18075272 1 mg 1mg
1 options
Product Code. 18075272
Brand: Tocris Bioscience™ 3015/1
Potent and selective PAR2 agonist
2-Furoyl-LIGRLO-amide is a potent and selective PAR2 receptor agonist (pD2 = 7.0). Causes a dose-dependent relaxation of murine femoral arteries.
TRUSTED_SUSTAINABILITY
Specifications
Purity 0.95
Quantity 1 mg