Learn More
α-Tocopherol Polyethylene Glycol Succinate, BLD Pharm™
Specifications
Specifications
| CAS | 9002-96-4 |
| Molecular Formula | (C2H4O)nC33H54O5 |
| Quantity | 1 g |
| SMILES | NONE |
| Chemical Name or Material | α-Tocopherol Polyethylene Glycol Succinate |