Learn More
α-Tocopherol phosphate, MedChemExpress
Description
α-Tocopherol phosphate is the compound demonstrating the highest vitamin E activity, which is available both in its natural form as RRR-alpha-tocopherol isolated from plant sources.
Specifications
Specifications
| Chemical Name or Material | α-Tocopherol phosphate |
| CAS | 38976-17-9 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C29H51O5P |
| Quantity | 10 mg |
| Synonym | alpha-Tocopherol phosphate TocP vitamin E phosphate |
| Solubility Information | DMSO : ≥ 100 mg/mL (195.81 mM) ∣H2O : 0.67 mg/mL (1.31 mM; Need ultrasonic) |
| SMILES | CC1=C(O[C@@](CCC[C@H](C)CCC[C@H](C)CCCC(C)C)(C)CC2)C2=C(C)C(OP(O)(O)=O)=C1C |
| Molecular Weight (g/mol) | 510.69 |
| Show More |