missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNIP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15104-1-AP-20UL
This item is not returnable.
View return policy
Description
Grasp65 is a structural protein of the golgi apparatus. It has been implicated in the stacking of golgi cisternae. Grasp65 is shown to have the properties to bind surfaces together in a mitotically regulated matter.Specifications
| TNIP1 | |
| Polyclonal | |
| Unconjugated | |
| TNIP1 | |
| ABIN 1, ABIN1, ABIN-1, HIV 1 Nef interacting protein, hVAN, KIAA0113, NAF1, Nef associated factor 1, Nip40 1, TNFAIP3 interacting protein 1, TNIP1, VAN | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10318, 57783 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q15025, Q9WUU8 | |
| Tnip1 | |
| TNIP1 Fusion Protein Ag1561 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction