missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ TNFRSF7 (Human) Recombinant Protein

Human TNFRSF7 full-length ORF ( AAH12160, 1 a.a. - 260 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00000939-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16142831

  • 4130.00 NOK / 10µg
Estimated Shipment: 01-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sequence: MARPHPWWLCVLGTLVGLSATPAPKSCPERHYWAQGKLCCQMCEPGTFLVKDCDQHRKAAQCDPCIPGVSFSPDHHTRPHCESCRHCNSGLLVRNCTITANAECACRNGWQCRDKECTECDPLPNPSLTARSSQALSPHPQPTHLPYVSEMLEARTAGHMQTLADFRQLPARTLSTHWPPQRSLCSSDFIRILVIFSGMFLVFTLAGALFLHQRRKYRSNKGESPVEPAEPCRYSCPREEEGSTIPIQEDYRKPEPACSP
Specifications

Specifications

AAH12160
CD27 molecule
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CD27
GST
939
Wheat germ expression system
10 μg
MGC20393, S152, T14, TNFRSF7, Tp55
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents
Certificates
Promotions

Promotions