missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMPRSS2 Antibody (CL12512), Novus Biologicals™
Mouse Monoclonal Antibody
3555.00 NOK - 5500.00 NOK
Specifications
| Antigen | TMPRSS2 |
|---|---|
| Clone | CL12512 |
| Dilution | Immunohistochemistry 1:10000 - 1:20000, Immunohistochemistry-Paraffin 1:10000 - 1:20000 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18640895
|
Novus Biologicals
NBP3-18573-100ul |
100 μg |
5500.00 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18631644
|
Novus Biologicals
NBP3-18573-25ul |
25 μg |
3555.00 NOK
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TMPRSS2 Monoclonal antibody specifically detects TMPRSS2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| TMPRSS2 | |
| Immunohistochemistry 1:10000 - 1:20000, Immunohistochemistry-Paraffin 1:10000 - 1:20000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 3.4.21, FLJ41954, NP_001128571.1, NP_001369649.1, NP_005647.3, PP9284, PRSS10, Serine protease 10, TMPRSS2, transmembrane protease serine 2, transmembrane protease, serine 2 | |
| This antibody was developed against a recombinant protein of O15393. Actual immunogen sequence is proprietary. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| CL12512 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Biology, Neuroscience, Prostate Cancer, Virology Bacteria and Parasites | |
| PBS, pH 7.2, 40% glycerol | |
| 7113 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title