missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMPRSS2 Antibody (1038105), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB107231P
This item is not returnable.
View return policy
Description
TMPRSS2 Monoclonal antibody specifically detects TMPRSS2 in Human samples. It is validated for Flow Cytometry
Specifications
| TMPRSS2 | |
| Monoclonal | |
| PE | |
| EC 3.4.21, FLJ41954, NP_001128571.1, NP_001369649.1, NP_005647.3, PP9284, PRSS10, Serine protease 10, TMPRSS2, transmembrane protease serine 2, transmembrane protease, serine 2 | |
| Human TMPRSS2 synthetic peptide, Accession # O15393 | |
| 0.1 mL | |
| Cancer, Cell Biology, Neuroscience, Prostate Cancer, Virology Bacteria and Parasites | |
| 7113 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry | |
| 1038105 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction