Learn More
TMPO/LAP2 Mouse anti-Human, Unconjugated, Clone: 3G4E1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67157-1-IG-150UL
Description
The TMPO/LAP2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TMPO/LAP2. This antibody recognizes human, rat, mouse antigen. The TMPO/LAP2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| TMPO/LAP2 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, ELISA | |
| 3G4E1 | |
| Unconjugated | |
| 150 μL | |
| 7112 | |
| IgG1 |