Learn More
TMPO/LAP2 Mouse anti-Human, Unconjugated, Clone: 3G4E1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67157-1-IG-20UL
 View more versions of this product
Product Code. 16809635
  • 1710.33 NOK / 20µL
Description
The TMPO/LAP2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TMPO/LAP2. This antibody recognizes human, rat, mouse antigen. The TMPO/LAP2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
TMPO/LAP2
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, ELISA
3G4E1
Unconjugated
20 μL
7112
IgG1