missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMOD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10145-1-AP-150UL
This item is not returnable.
View return policy
Description
TMOD1 gene encodes a member of the tropomodulin family. The encoded protein is an actin-capping protein that regulates tropomyosin by binding to its N-terminus, inhibiting depolymerization and elongation of the pointed end of actin filaments and thereby influencing the structure of the erythrocyte membrane skeleton. Multiple transcript variants encoding the same protein have been found for this gene.Specifications
| TMOD1 | |
| Polyclonal | |
| Unconjugated | |
| TMOD1 | |
| D9S57E, E Tmod, Erythrocyte tropomodulin, ETMOD, TMOD, TMOD1, tropomodulin 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 21916, 25566, 7111 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P28289, P49813, P70567 | |
| Tmod1 | |
| TMOD1 Fusion Protein Ag0192 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction