missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TMF1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16591022
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16591022 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16591022 Supplier Invitrogen™ Supplier No. PA598201

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TMF1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Adapter protein that may provide indirect link between the endocytic membrane traffic and the actin assembly machinery. May regulate the formation of clathrin-coated vesicles. Isoform 1 could be involved in brain-specific synaptic vesicle recycling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMF1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3.73 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene TMF1
Gene Accession No. P82094
Gene Alias 7030402D04Rik; Androgen receptor coactivator 160 kDa protein; androgen receptor-associated protein of 160 kDa; ARA160; Gm153; TATA element modulatory factor; TATA element modulatory factor 1; TMF; TMF1; Tmf1
Gene Symbols TMF1
Host Species Rabbit
Immunogen Recombinant Human TATA element modulatory factor protein (878-986AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7110
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.