missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TMEM68 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15869673
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15869673 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15869673 Supplier Invitrogen™ Supplier No. PA571266

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Dog: 92%; Guinea Pig: 77%; Horse: 79%; Human: 100%; Mouse: 79%; Pig: 85%; Rabbit: 85%; Rat: 85%.

TMEM68 gene ontology annotations related to this gene include transferase activity, transferring acyl groups.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM68
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TMEM68
Gene Accession No. Q96MH6
Gene Alias 2010300G19Rik; AA408325; AW123402; RGD1309006; Tmem68; Transmembrane protein 68; Transmembrane protein 68-like protein
Gene Symbols TMEM68
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human TMEM68.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 137695
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.