missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM214 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20125-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Transmembrane protein 214 (TMEM214) is a 77kDa membrane protein which is widely expressed at high level. The gene encoding this protein maps to chromosome 2, at 2p23.3 TMEM214 interacts with LSM1, an SM-like protein, forming a stable heteromer present in tri-snRNP particles, which are important for pre-mRNA splicing. The detailed function of TMEM214 is still unknown, but a recent study showed that TMEM214 may be used to explore the contribution of human host factors of some infectious diseases such as Dengue fever, West Nile fever and yellow fever. Multiple isoforms of TMEM214 are known to exist.Specifications
TMEM214 | |
Polyclonal | |
Unconjugated | |
TMEM214 | |
TMEM214, transmembrane protein 214 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
362711, 54867, 68796 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
A1L1L2, Q6NUQ4, Q8BM55 | |
Tmem214 | |
TMEM214 Fusion Protein Ag13884 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |