missing translation for 'onlineSavingsMsg'
Learn More

TMEM214 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 20125-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16899614

  • 1611.84 NOK / 20µL
Estimated Shipment: 23-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Transmembrane protein 214 (TMEM214) is a 77kDa membrane protein which is widely expressed at high level. The gene encoding this protein maps to chromosome 2, at 2p23.3 TMEM214 interacts with LSM1, an SM-like protein, forming a stable heteromer present in tri-snRNP particles, which are important for pre-mRNA splicing. The detailed function of TMEM214 is still unknown, but a recent study showed that TMEM214 may be used to explore the contribution of human host factors of some infectious diseases such as Dengue fever, West Nile fever and yellow fever. Multiple isoforms of TMEM214 are known to exist.
Specifications

Specifications

TMEM214
Polyclonal
Unconjugated
TMEM214
TMEM214, transmembrane protein 214
Rabbit
Antigen Affinity Chromatography
RUO
362711, 54867, 68796
-20°C
Liquid
Immunofluorescence, Immunocytochemistry, Western Blot
0.17 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
A1L1L2, Q6NUQ4, Q8BM55
Tmem214
TMEM214 Fusion Protein Ag13884
20 μL
Primary
Mouse, Human, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers