missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM175 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19925-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
TMEM175 gene ontology annotations related to this gene include potassium channel activity.Specifications
TMEM175 | |
Polyclonal | |
Unconjugated | |
TMEM175 | |
TMEM175, transmembrane protein 175 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
305623, 72392, 84286 | |
-20°C | |
Liquid |
Western Blot | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6AY05, Q9BSA9, Q9CXY1 | |
Tmem175 | |
TMEM175 Fusion Protein Ag13890 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |