missing translation for 'onlineSavingsMsg'
Learn More

TMEM175 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 19925-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16889394

  • 4593.76 NOK / 150µL
Estimated Shipment: 21-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

TMEM175 gene ontology annotations related to this gene include potassium channel activity.
Specifications

Specifications

TMEM175
Polyclonal
Unconjugated
TMEM175
TMEM175, transmembrane protein 175
Rabbit
Antigen Affinity Chromatography
RUO
305623, 72392, 84286
-20°C
Liquid
Western Blot
0.24 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6AY05, Q9BSA9, Q9CXY1
Tmem175
TMEM175 Fusion Protein Ag13890
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers