Learn More
TMEM173/STING Rabbit, anti-Human, Unconjugated, Clone: 7K7, Proteintech
Shop All Proteintech ProductsDescription
TMEM173, also named as ERIS, MITA, STING and MPYS, is a facilitator of innate immune signaling that promotes the production of type I IFN (IFN-alpha and IFN-beta). TMEM173 mediates death signaling via activation of the extracellular signal-regulated kinase (ERK) pathway.
Specifications
Specifications
| Antigen | TMEM173/STING |
| Applications | Western Blot, Immunohistochemistry, ELISA |
| Classification | Monoclonal |
| Clone | 7K7 |
| Concentration | 1000 μg/mL |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Quantity | 100 μL |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 340061 |
| Show More |