missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM168 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22527-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
TMEM168 is a multi-pass membrane protein. It belongs to the TMEM168 family. The exact function of TMEM168 remains unknown.Specifications
TMEM168 | |
Polyclonal | |
Unconjugated | |
TMEM168 | |
TMEM168, transmembrane protein 168 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
101118, 312135, 64418 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.55 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5PQM0, Q91VX9, Q9H0V1 | |
Tmem168 | |
TMEM168 Fusion Protein Ag18337 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |