Learn More
TMEM165 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20485-1-AP-150UL
 View more versions of this product
Product Code. 16800035
  • 4836.10 NOK / 150µL
Description
The function of TM165 remains unknown.
Specifications
Specifications
TMEM165
Polyclonal
Unconjugated
TMEM165
TMEM165, TMPT27, TPARL, transmembrane protein 165, Transmembrane protein PT27, Transmembrane protein TPARL
Rabbit
Antigen Affinity Chromatography
RUO
21982, 364137, 55858
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P52875, Q4V899, Q9HC07
Tmem165
TMEM165 Fusion Protein Ag14362
150 μL
Primary
Human, Mouse, Rat