Learn More
TMEM11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16564-1-AP-20UL
This item is not returnable.
View return policy
Description
Plays a role in mitochondrial morphogenesis.Specifications
| TMEM11 | |
| Polyclonal | |
| Unconjugated | |
| TMEM11 | |
| C17orf35, PM1, PMI, Protein PM1, TMEM11, transmembrane protein 11 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 216821, 303196, 8834 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| B0BN86, P17152, Q8BK08 | |
| TMEM11 | |
| TMEM11 Fusion Protein Ag9839 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |