missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TMA16 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17199273
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17199273 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17199273 Supplier Invitrogen™ Supplier No. PA5111360

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: KALRLNLVGE KLQWFQNHLD PQKKRYSKKD ACELIERYLN RFSSELEQIE LHNSIRDRQG RRHCSRETVI KQTMER Highest antigen sequence indentity to the following orthologs: Rat - 84%, Mouse - 82%.

TMA16 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMA16
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TMA16
Gene Accession No. Q96EY4
Gene Alias 1810029B16Rik; C4orf43; RGD1305222; TMA16; translation machinery associated 16 homolog; translation machinery associated 16 homolog (S. cerevisiae); translation machinery-associated protein 16; UPF0534 protein C4orf43; UPF0534 protein C4orf43 homolog
Gene Symbols TMA16
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human TMA16. Recombinant protein control fragment (Product #RP-106410).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55319
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.