Learn More
TM9SF4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25595-1-AP-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16884155
  • 1281.92 NOK / 20µL
Estimated Shipment: 20-12-2022
Description
TM9SF4 is a protein coding gene.
Specifications
Specifications
TM9SF4
Polyclonal
Unconjugated
TM9SF4
KIAA0255, TM9SF4
Rabbit
Antigen Affinity Chromatography
RUO
9777
-20°C
Liquid
Western Blot
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q92544
TM9SF4
TM9SF4 Fusion Protein Ag22335