missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR2 Mouse anti-Human, Clone: 1C1D3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66645-1-IG-20UL
This item is not returnable.
View return policy
Description
The TLR2 protein is a member of the Toll-like receptor (TLR) family which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved from Drosophila to humans and share structural and functional similarities. They recognize pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents, and mediate the production of cytokines necessary for the development of effective immunity. The various TLRs exhibit different patterns of expression. This protein is expressed most abundantly in peripheral blood leukocytes, and mediates host response to Gram-positive bacteria and yeast via stimulation of NF-kappaB.Specifications
| TLR2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O60603 | |
| TLR2 | |
| TLR2 Fusion Protein Ag25626 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 1C1D3 | |
| Unconjugated | |
| TLR2 | |
| CD282, TIL4, TLR2, toll like receptor 2 | |
| Mouse | |
| Protein A | |
| RUO | |
| 7097 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction