Learn More
MilliporeSigma™ TLC Silica Gel 60 Plate with F254 Coating
Description
- Combination of silica gel 60, addition of polymeric binder
- Adherent hard surface that will not crack or blister
- Size: 10 x 20cm
Specifications
Specifications
| Backing Material | Aluminum Support |
| Color | White |
| Quantity | 25 Units/Pk. |
| Type | Silica Gel |
| For Use With (Application) | Thin Layer Chromatography |
| Storage Requirements | Room temperature |