Learn More
Macherey-Nagel™ TLC Polyester Sheets
Product Code. 10378501 Shop All Macherey-Nagel Products
Click to view available options
For Use With (Equipment):
All TLC Micro Sets
Unit Size:
Pack of 200
Product Code. 10378501
Brand: Macherey-Nagel™ 814025
Only null left
Add to basket
Add to basket
Materials kit for scientific education. Macherey Nagel™ Polygram™ TLC Polyester Sheets provide basic equipment for the individual study of further thin layer chromatographic experiments and are components of the TLC micro-set M.
  • This kit is prerequisite for the separations with kits F1, F2 and F3. In addition, it serves as basic equipment for the individual study of further thin layer chromatographic experiments.
Specifications
Technique TLC Micro Sets
Quantity 200 Pack
Type TLC Polyester Sheets
For Use With (Equipment) All TLC Micro Sets