missing translation for 'onlineSavingsMsg'
Learn More

Tivantinib, MedChemExpress

Product Code. p-200096338
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16461316

Brand: Medchem Express HY50686

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tivantinib is a highly selective c-Met tyrosine kinase inhibitor with a Ki of 355 nM.

Tivantinib is a highly selective c-Met tyrosine kinase inhibitor with a Ki of 355 nM.

Specifications

Chemical Name or Material Tivantinib
CAS 905854-02-6
Color Brown
Purity Grade Notes Research
Molecular Formula C23H19N3O2
Quantity 200 mg
Synonym ARQ 197
Solubility Information DMSO : ≥ 100 mg/mL (270.69 mM)
SMILES O=C(NC1=O)[C@@H](C2=CN3C4=C(C=CC=C42)CCC3)[C@@H]1C5=CNC6=C5C=CC=C6
Molecular Weight (g/mol) 369.42
Formula Weight 369.42
Percent Purity 96.99%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.