missing translation for 'onlineSavingsMsg'
Learn More

Titriplex™ VI, Supelco™

Product Code. 15827680
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15827680 100 g 100g
15817680 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15827680 Supplier Supelco Supplier No. 1084350100
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For analysis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-42-5

Specifications

Melting Point 240°C to 245°C (decomposes)
MDL Number MFCD00004291
Quantity 100 g
Synonym Titriplex™ VI, 3,6-Dioxaoctamethylenedinitrilotetraacetic acid, Ethylenedioxybis(ethylenenitrilo)tetraacetic acid, Bis(aminoethyl)glycolether-N,N,N,N′-tetraacetic acid, Ethylene glycol-bis (β-amino ethyl ether)-N,N′-tetraacetic acid, Titriplex™ VI
Molecular Weight (g/mol) 380.35
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.