Learn More
SI Analytics™ Titration Vessel
Product Code. 10672832 Shop All SI Analytics Products
Change view
Click to view available options
Volume (Metric):
3 to 10 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10672832 3 to 10 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10672832 Supplier SI Analytics™ Supplier No. 285213499
  • ONLINE EXCLUSIVE PRICE
    20555.00 NOK
    19309.03 NOK / Pack of 1
    Save 1245.97 NOK
    6% Off
 Restricted Item
Estimated Shipment: 04-01-2027
SI Analytics™ Titration Vessel is a temperature controlled vessel that features a double jacket with 5 openings.
  • Temperature-controlled titration vessel
  • Double jacket
  • 4 openings NS 14.5 and 1 opening NS 7.5
  • Includes a micro-titration tip with PTFE insert and gas supply tube
  • Micro design
TRUSTED_SUSTAINABILITY
Specifications
Type Titration Vessel
Includes Micro titration tip with PTFE insert and gas supply tube
Size 4 x NS 14.5, 1 x NS 7.5
Volume (Metric) 3 to 10 mL