Learn More
SI Analytics™ Titration Tip
Product Code. 10253701 Shop All SI Analytics Products
Change view
Click to view available options
Length (Metric):
120 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10253701 120 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10253701 Supplier SI Analytics™ Supplier No. 285213717
  • ONLINE EXCLUSIVE PRICE
    945.00 NOK
    790.29 NOK / Pack of 1
    Save 154.71 NOK
    16% Off
Estimated Shipment: 14-12-2026
SI Analytics™ Titration Tip is a connector for 20 mL and 50 mL burette cylinders.
  • 120 mm length
  • Capillary Ø 0.8 mm
  • For 20 mL and 50 mL burette cylinder
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 0.8 mm capillary
Length (Metric) 120 mm