Learn More
Heidolph™ Titramax 100 Shaker
Product Code. 10324441 Shop All Heidolph Instruments Products
Change view
Click to view available options
Orbit:
1.5 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Orbit Unit Size
10324441 1.5 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10324441 Supplier Heidolph™ Supplier No. 5441120000
Discount Lab Supplies
Use promo code 28170
View all promotions
Ideal for use in science, research, and other laboratories, this Titramax 100 Shaker is a compact and space-saving unit that accepts load capacities of 2 kg and space for 4 microtiter plates.
  • Titramax 100 is a compact and space-saving unit that accepts load capacities of 2 kg and space for 4 microtiter plates
  • The 1.5 mm vibration orbit performs gentle mixing with best results using small sample volumes
  • Set and continuously adjust the variable speed on the analog control knob from 150 to 1,350 rpm
  • An analog process timer allows for unattended operation and can be set from 1 to 120 minutes
  • When the set time has elapsed an acoustic alarm will sound and the operation stops
TRUSTED_SUSTAINABILITY
Specifications
Orbit 1.5 mm
Load Bearing Capacity (Metric) 2 kg
Speed Range 150 to 1350 rpm
Electrical Requirements 230 V, 50 Hz, 15 W
Temperature Range (Metric) 0°C to 50°C
Holds 4 Microtiter Plates
Load Bearing Capacity (English) 4.4 lb.
Temperature Range (English) 32°F to 122°F
Dimensions (LxW) Platform 220 x 220 mm
Dimensions (L x W x H) 245 x 310 x 125 mm
Weight (English) 12.1 lb.
Weight (Metric) 5.5 kg
Model Titramax 100
Timer Range 0 to 120 min., Continuous