Learn More
Heidolph™ Titramax 101 Shaker/Mixer
Product Code. 10101031 Shop All Heidolph Instruments Products
Change view
Click to view available options
Orbit:
3 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Orbit Unit Size
10101031 3 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10101031 Supplier Heidolph™ Supplier No. 5441130000
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
 Restricted Item
Estimated Shipment: 11-12-2026
Titramax 101 Shaker/Mixer is a compact and space-saving unit that accepts load capacities of 2 kg and space for four microtiter plates.
  • 3 mm vibration orbit performs gentle mixing with best results using larger sample volumes
  • Settable and continuously adjustable speed from 150 to 1,350rpm
  • Analog process timer can be set form 1 to 120 minutes for unattended operation
  • Acoustic alarm and shut off at the end of timer operation
TRUSTED_SUSTAINABILITY
Specifications
Orbit 3 mm
Load Bearing Capacity (Metric) 2 kg
Speed Range 150 to 1350 rpm
Wattage 31 W
Product Type Shaker Mixer
Load Bearing Capacity (English) 4.4 lb.
Certifications/Compliance Protection class DIN EN 60529, IP 30
Model Titramax 101
Timer Range 1 to 120 min.