Learn More
MP Biomedicals™ Titertek™ Finnpipette™ Multistepper Pipet Tips
Shop All MP Biomedicals Products
Click to view available options
Sterility:
Non-sterile
Sterile
Format:
Bulk
Rack
Unit Size:
Pack of 1000
Pack of 960
Description
Specifically for the Titertek™/Labsystems 8-channel Multistepper pipette
- Made from virgin polypropylene material
- Comes with robot-compatible (large-micron) hydrophobic filters to eliminate instrument contamination
- 96 tips per rack
- Volume: 1500μL
Specifications
Specifications
| Volume (Metric) | 1500 μL |
| Sterility | Sterile |
| Color | Natural |
| Format | Rack |
| Material | Polypropylene |