missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium, Ti 8-1-1, R54810, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Titanium, Ti 8-1-1, R54810, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Chromium/Zirconium/Copper/Niobium/Nitrogen/Hafnium/Manganese/Tin/Carbon/Silicon/Iron/Oxygen/Molybdenum/Vanadium/Aluminium/Magnesium/Cobalt/Boron/Yttrium/Palladium/Tantalum/Nickel/Tungsten/Phosphorus/Ruthenium/Titanium |
| CAS | 7440-32-6 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Chromium 0.0014 %/Zirconium 0.002 %/Copper 0.0023 %/Niobium 0.004 %/Nitrogen 0.006 %/Hafnium 0.007 %/Manganese 0.0071 %/Tin 0.008 %/Carbon 0.014 %/Silicon 0.025 %/Iron 0.071 %/Oxygen 0.09 %/Molybdenum 0.98 %/Vanadium 1.03 %/Aluminium 7.86 %/Magnesium (<0.0001) %/Cobalt (<0.0005) %/Boron (<0.001) %/Yttrium (<0.001) %/Palladium (<0.005) %/Tantalum (<0.005) %/Nickel (0.001) %/Tungsten (0.001) %/Phosphorus (0.003) %/Ruthenium (0.004) %/Titanium (89.9) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction