Learn More
Thermo Scientific™ Titanium Buckets for SureSpin™ 632 rotor
Product Code. 16644884
Change view
Click to view available options
Capacity:
17 mL
38.5 mL
Unit Size:
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity unitSize
16644884 38.5 mL Pack of 6
16634884 17 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16644884 Supplier Thermo Scientific™ Supplier No. 75003037
  • ONLINE EXCLUSIVE PRICE
    249050.00 NOK
    242164.74 NOK / Pack of 6
    Save 6885.26 NOK
    2% Off
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
For use with SureSpin™ 632 rotor
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) SureSpin 632
Capacity 38.5 mL
Material Titanium
Quantity 6/Pk.