Learn More
Epredia™ TissueLoc™ Cassette IV Slotted Cassettes
Shop All Epredia Products
Click to view available options
Color:
Beige
Blue
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
Unit Size:
Pack of 1000
Description
Printing
- Surface finish and reinforced print faces are engineered to optimize print quality
- Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
- Cassette design and flow characteristics tested to ensure efficient reagent exchange
- Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
- Precision engineering for solid connection to microtomes
Archiving
- Cassette surface texture optimizes scannability for archiving
Specifications
Specifications
| Pore Style | Slots |
| Closure Included | Yes |
| Packaging Type | Loose Packed |
| Quantity | 1000/Cs. |
| Closure Type | Separate Lid, Back Hinge |
| PrintMate Compatible | No |
| Color | Tan |
| Product Type | Slotted Cassette |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion