Learn More
Epredia™ TissueLoc™ Cassette IV Slotted Cassettes
Product Code. 16324176 Shop All Epredia Products
Click to view available options
Color:
Beige
Blue
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
Unit Size:
Pack of 1000
Product Code. 16324176
Brand: Epredia™ B851954GN
For routine tissue samples, cassettes have a back-edge lid hinge with loose-packed lids that are available in 12 different color varieties
Printing
  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
  • Precision engineering for solid connection to microtomes
Archiving
  • Cassette surface texture optimizes scannability for archiving
TRUSTED_SUSTAINABILITY
Specifications
Pore Style Slots
Closure Included Yes
Packaging Type Loose Packed
Quantity 1000/Cs.
Closure Type Separate Lid, Back Hinge
PrintMate Compatible No
Color Green
Product Type Slotted Cassette