Learn More
Epredia™ TissueLoc™ Cassette IV Slotted Cassettes
Product Code. 16304176 Shop All Epredia Products
Change view
Click to view available options
Color:
Beige
Blue
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
Unit Size:
Pack of 1000
Product Code. Color unitSize
16304176 Beige Pack of 1000
16314176 Blue Pack of 1000
16324176 Green Pack of 1000
16344176 Lavender Pack of 1000
16354176 Light Green Pack of 1000
16364176 Lilac Pack of 1000
16374176 Orange Pack of 1000
16384176 Pink Pack of 1000
16394176 Tan Pack of 1000
16304186 White Pack of 1000
16314186 Yellow Pack of 1000
11 options
Product Code. 16304176
Brand: Epredia™ B851954BE
For routine tissue samples, cassettes have a back-edge lid hinge with loose-packed lids that are available in 12 different color varieties
Printing
  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
  • Precision engineering for solid connection to microtomes
Archiving
  • Cassette surface texture optimizes scannability for archiving
TRUSTED_SUSTAINABILITY
Specifications
Pore Style Slots
Closure Included Yes
Packaging Type Loose Packed
Quantity 1000/Cs.
Closure Type Separate Lid, Back Hinge
PrintMate Compatible No
Color Beige
Product Type Slotted Cassette